Skip to Content
Merck
Manganese(IV) oxide ReagentPlus®, ≥99%

Manganese(IV) oxide

Manganese(IV) oxide ReagentPlus®, ≥99%
Synonym(s):
Manganese dioxide, Manganese(IV) oxide
Empirical Formula (Hill Notation):
MnO2
CAS Number:
Molecular Weight:
86.94

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Particle Size (µm)
Reagent Type
Showing 1-5 of 5
Compare
Product Number
Description
Pricing