Mafenide Related Compound A
Synonym(s):
4-Formylbenzenesulfonamide
Empirical Formula (Hill Notation):
C7H7NO3S
CAS Number:
Molecular Weight:
185.20
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2