
(-)-Matairesinol

Synonym(s):
Matairesinol, (αR,βR)-α,β-Bis(4-hydroxy-3-methoxybenzyl)butyrolactone
Empirical Formula (Hill Notation):
C20H22O6
CAS Number:
Molecular Weight:
358.39
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing