L-Kynurenine
Synonym(s):
β-Anthraniloyl-L-alanine, L-2-Amino-4-(2-aminophenyl)-4-oxobutanoic acid
Empirical Formula (Hill Notation):
C10H12N2O3
CAS Number:
Molecular Weight:
208.21
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Optical Isomer
Showing 1-2 of 2
Compare
Product Number
Description
Pricing