Lithium manganese oxide
Synonym(s):
Lithium manganese(III,IV) oxide, LMO
Linear Formula:
LiMn2O4
CAS Number:
Molecular Weight:
180.81
Products
Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Application
Particle Size (µm)
Showing 1-1 of 1
Compare
Product Number
Description
Pricing