Linalyl acetate
Synonym(s):
3,7-Dimethyl-1,6-octadien-3-yl acetate, Bergamol
Linear Formula:
CH3CO2C(CH=CH2)(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
196.29
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing