Skip to Content
Merck
L-Glutamic acid dimethyl ester hydrochloride ≥99.0% (anhydrous basis material, AT)

L-Glutamic acid dimethyl ester hydrochloride

L-Glutamic acid dimethyl ester hydrochloride ≥99.0% (anhydrous basis material, AT)
Linear Formula:
CH3COOCH2CH2CH(NH2)COOCH3 · HCl
CAS Number:
Molecular Weight:
211.64
EC Number:

Products

Brand
Formula Weight
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Special Grade
Showing 1-1 of 1