![Levothyroxine sodium European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/183/201/ec3a05dd-717f-432a-b4d3-88bdb4ba8dda/640/ec3a05dd-717f-432a-b4d3-88bdb4ba8dda.png)
Levothyroxine sodium
![Levothyroxine sodium European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/183/201/ec3a05dd-717f-432a-b4d3-88bdb4ba8dda/640/ec3a05dd-717f-432a-b4d3-88bdb4ba8dda.png)
Synonym(s):
Levothyroxine sodium hydrate, 3,3′,5,5′-Tetraiodo-L-thyronine monosodium salt hydrate, L-Thyroxin monosodium salt hydrate, Sodium O-(4-hydroxy-3,5-diiodophenyl)-3,5-diiodo-L-tyrosine hydrate
Empirical Formula (Hill Notation):
C15H10I4NNaO4 · xH2O
CAS Number:
Molecular Weight:
798.85 (anhydrous basis)
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing