Skip to Content
BE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
L-Asparagine monohydrate
Synonym(s):
(
S
)-(+)-2-Aminosuccinamic acid, (
S
)-2-Aminosuccinic acid 4-amide, L-Aspartic acid 4-amide
Linear Formula:
NH
2
COCH
2
CH(NH
2
)COOH · H
2
O
CAS Number:
5794-13-8
Molecular Weight:
150.13
EC Number:
200-735-9
Products
Product Category
amino acids (3)
Brand
SAFC (1)
Sigma-Aldrich (3)
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Functional Group
Application
Optical Isomer
Amino Acid
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
A7094
L-Asparagine monohydrate,
≥98.0%, suitable for cell culture, BioReagent, non-animal source
Expand
View Pricing
A8381
L-Asparagine monohydrate,
≥99% (TLC)
Expand
View Pricing
A4284
L-Asparagine monohydrate,
suitable for cell culture, BioReagent
Expand
View Pricing
RES10070-A7
L-Asparagine monohydrate,
Expand
View Pricing