![L-Arginine-7-amido-4-methylcoumarin hydrochloride cathepsin H substrate](/deepweb/assets/sigmaaldrich/product/structures/397/524/d174b0dc-5cbe-46fb-b548-98437d03c4e0/640/d174b0dc-5cbe-46fb-b548-98437d03c4e0.png)
L-Arginine-7-amido-4-methylcoumarin hydrochloride
![L-Arginine-7-amido-4-methylcoumarin hydrochloride cathepsin H substrate](/deepweb/assets/sigmaaldrich/product/structures/397/524/d174b0dc-5cbe-46fb-b548-98437d03c4e0/640/d174b0dc-5cbe-46fb-b548-98437d03c4e0.png)
Empirical Formula (Hill Notation):
C16H21N5O3
CAS Number:
Molecular Weight:
331.37
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing