Lanoconazole
Synonym(s):
2-[4-(2-Chlorophenyl)-1,3-dithiolan-2-ylidene]-2-imidazol-1-yl-acetonitrile
Empirical Formula (Hill Notation):
C14H10ClN3S2
CAS Number:
Molecular Weight:
319.83
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing