Ketorolac Related Compound A
Synonym(s):
5-Benzoyl-N-(1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)-2,3-dihydro-1H-pyrrolizine-1-carboxamide
Empirical Formula (Hill Notation):
C19H22N2O5
CAS Number:
Molecular Weight:
358.39
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2