JohnPhos
Synonym(s):
(2-Biphenyl)di-tert-butylphosphine, (2-Biphenylyl)di-tert-butylphosphine, 2-(Di-tert-butylphosphino)biphenyl
Linear Formula:
C6H5C6H4P[C(CH3)3]2
CAS Number:
Molecular Weight:
298.40
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing