Skip to Content
Merck
Iron(III) sulfate hydrate BioReagent, suitable for plant cell culture

Iron(III) sulfate hydrate

Iron(III) sulfate hydrate BioReagent, suitable for plant cell culture
Synonym(s):
Ferric sulfate hydrate
Linear Formula:
Fe2(SO4)3 · xH2O
CAS Number:
Molecular Weight:
399.88 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Application
Reagent Type
Core
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing