Skip to Content
Merck
vibrant-m

Insulin porcine for system suitability

vibrant-m
Synonym(s):
Insulin from porcine pancreas, Insulin from hog pancreas
Empirical Formula (Hill Notation):
C256H381N65O76S6
CAS Number:
Molecular Weight:
5777.54

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1