![InP/ZnS quantum dots stabilized with oleylamine ligands, fluorescence λem 560 nm, 5 mg/mL in toluene](/deepweb/assets/sigmaaldrich/product/structures/371/579/2632e7e2-e515-4eef-9402-531b205c83d8/640/2632e7e2-e515-4eef-9402-531b205c83d8.png)
InP/ZnS quantum dots
![InP/ZnS quantum dots stabilized with oleylamine ligands, fluorescence λem 560 nm, 5 mg/mL in toluene](/deepweb/assets/sigmaaldrich/product/structures/371/579/2632e7e2-e515-4eef-9402-531b205c83d8/640/2632e7e2-e515-4eef-9402-531b205c83d8.png)
Synonym(s):
Cadmium free core shell quantum dots, Cadmium free quantum dots, Core/Shell heavy metal free quantum dots, Fluorescent nanocrystals
Linear Formula:
InP/ZnS
Products
Product Category
Brand
Physical Form
Quality Segment
Showing 1-5 of 5
Compare
Product Number
Description
Pricing