Indium oxide coated PET
Synonym(s):
IO-PET, Polyethylene terephthalate film, indium oxide coated
Empirical Formula (Hill Notation):
In2O3
Molecular Weight:
277.63
Products
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing