Skip to Content
Merck
Indaziflam PESTANAL®, analytical standard

Indaziflam

Indaziflam PESTANAL®, analytical standard
Synonym(s):
2-Amino-4-[(1R,2S)-2,6-dimethyl-indan-1-ylamino]-6-(1-fluoroethyl)-1,3,5-triazine, N-[(1R,2S)-2,3-Dihydro-2,6-dimethyl-1H-inden-1-yl]-6-[(1RS)-1-fluoroethyl]-1,3,5-triazine-2,4-diamine
Empirical Formula (Hill Notation):
C16H20FN5
CAS Number:
Molecular Weight:
301.36

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing