![Imazamox PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/328/597/e403644e-fe7b-421c-afa6-a9553d72e360/640/e403644e-fe7b-421c-afa6-a9553d72e360.png)
Imazamox
![Imazamox PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/328/597/e403644e-fe7b-421c-afa6-a9553d72e360/640/e403644e-fe7b-421c-afa6-a9553d72e360.png)
Synonym(s):
2-(4-Isopropyl-4-methyl-5-oxo-2-imidazolin-2-yl)-5-methoxymethylnicotinic acid, 2-[4,5-Dihydro-4-methyl-4-(1-methylethyl)-5-oxo-1H-imidazol-2-yl]-5-(methoxymethyl)-3-pyridinecarboxylic acid
Empirical Formula (Hill Notation):
C15H19N3O4
CAS Number:
Molecular Weight:
305.33
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2