Hydrogen chloride solution
Synonym(s):
Hydrogen chloride – ethanol solution, Hydrochloric acid in ethanol, Hydrochloric acid solution, Hydrogen chloride solution
Linear Formula:
HCl · CH3CH2OH
CAS Number:
Molecular Weight:
36.46
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Quality Segment
Application
Package Size
Reaction Type
Reagent Type
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing