
Hydrastine hydrochloride

Synonym(s):
(1R,9S)-β-Hydrastine hydrochloride
Empirical Formula (Hill Notation):
C21H21NO6 · HCl
CAS Number:
Molecular Weight:
419.86
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing