Skip to Content
Merck
(Heptafluoropropyl)trimethylsilane 97%

(Heptafluoropropyl)trimethylsilane

(Heptafluoropropyl)trimethylsilane 97%
Synonym(s):
1-(Trimethylsilyl)heptafluoropropane
Linear Formula:
CF3CF2CF2Si(CH3)3
CAS Number:
Molecular Weight:
242.21

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing