![GYKI 52466 hydrochloride solid](/deepweb/assets/sigmaaldrich/product/structures/171/590/c7df5eb5-8120-4d66-a380-5cdafd2cf05a/640/c7df5eb5-8120-4d66-a380-5cdafd2cf05a.png)
GYKI 52466 hydrochloride
![GYKI 52466 hydrochloride solid](/deepweb/assets/sigmaaldrich/product/structures/171/590/c7df5eb5-8120-4d66-a380-5cdafd2cf05a/640/c7df5eb5-8120-4d66-a380-5cdafd2cf05a.png)
Synonym(s):
1-(4-Aminophenyl)-4-methyl-7,8-methylenedioxy-5H-2,3-benzodiazepine hydrochloride
Empirical Formula (Hill Notation):
C17H15N3O2 · HCl
CAS Number:
Molecular Weight:
329.78
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing