Skip to Content
BE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gold-iron alloy, Au99.93Fe0.07
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
GF43041002
Gold-iron alloy, Au99.93Fe0.07,
0.1m wire, 0.025mm diameter, annealed
Expand
View Pricing
GF32219298
Gold-iron alloy, Au99.93Fe0.07,
0.05m wire, 0.025mm diameter, annealed
Expand
View Pricing
GF24715425
Gold-iron alloy, Au99.93Fe0.07,
2m insulated wire, 0.1mm conductor diameter, 0.01mm insulation thickness, ptfe (polytetrafluoroethylene) insulation
Expand
View Pricing
GF43923539
Gold-iron alloy, Au99.93Fe0.07,
0.5m insulated wire, 0.1mm conductor diameter, 0.01mm insulation thickness, ptfe (polytetrafluoroethylene) insulation
Expand
View Pricing
GF16727418
Gold-iron alloy, Au99.93Fe0.07,
0.2m insulated wire, 0.1mm conductor diameter, 0.01mm insulation thickness, ptfe (polytetrafluoroethylene) insulation
Expand
View Pricing
GF31674955
Gold-iron alloy, Au99.93Fe0.07,
1m insulated wire, 0.1mm conductor diameter, 0.01mm insulation thickness, ptfe (polytetrafluoroethylene) insulation
Expand
View Pricing
GF86855114
Gold-iron alloy, Au99.93Fe0.07,
0.5m wire, 0.050mm diameter, annealed
Expand
View Pricing
GF31160517
Gold-iron alloy, Au99.93Fe0.07,
0.2m wire, 0.050mm diameter, annealed
Expand
View Pricing
GF06918116
Gold-iron alloy, Au99.93Fe0.07,
0.05m wire, 0.050mm diameter, annealed
Expand
View Pricing
GF32441363
Gold-iron alloy, Au99.93Fe0.07,
0.1m insulated wire, 0.1mm conductor diameter, 0.01mm insulation thickness, ptfe (polytetrafluoroethylene) insulation
Expand
View Pricing
GF15774060
Gold-iron alloy, Au99.93Fe0.07,
0.5m wire, 0.025mm diameter, annealed
Expand
View Pricing