Skip to Content
Merck
Glycidyltrimethylammonium chloride technical, ≥90% (calc. based on dry substance, AT)

Glycidyltrimethylammonium chloride

Glycidyltrimethylammonium chloride technical, ≥90% (calc. based on dry substance, AT)
Synonym(s):
(2,3-Epoxypropyl)trimethylammonium chloride
Empirical Formula (Hill Notation):
C6H14ClNO
CAS Number:
Molecular Weight:
151.63

Products

Brand
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1