Skip to Content
Merck
Glycidyl hexadecyl ether technical grade

Glycidyl hexadecyl ether

Glycidyl hexadecyl ether technical grade
Synonym(s):
2,3-Epoxypropyl hexadecyl ether
Empirical Formula (Hill Notation):
C19H38O2
CAS Number:
Molecular Weight:
298.50
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Monomer Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing