Skip to Content
Merck
Glycerol diglycidyl ether technical grade

Glycerol diglycidyl ether

Glycerol diglycidyl ether technical grade
Empirical Formula (Hill Notation):
C9H16O5
CAS Number:
Molecular Weight:
204.22

Products

Brand
Formula Weight
Markush Class
Markush Group
Monomer Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing