Skip to Content
Merck
Gly-Arg-Gly-Asp-Ser ≥97% (HPLC)

Gly-Arg-Gly-Asp-Ser

Gly-Arg-Gly-Asp-Ser ≥97% (HPLC)
Empirical Formula (Hill Notation):
C17H30N8O9
CAS Number:
Molecular Weight:
490.47

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Chemical Composition
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing