![Glucosamine for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/457/151/aa4b7f67-13db-47a5-b6aa-71855f06a38a/640/aa4b7f67-13db-47a5-b6aa-71855f06a38a.png)
Glucosamine for system suitability
![Glucosamine for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/457/151/aa4b7f67-13db-47a5-b6aa-71855f06a38a/640/aa4b7f67-13db-47a5-b6aa-71855f06a38a.png)
Synonym(s):
D-(+)-Glucosamine hydrochloride, 2-Amino-2-deoxy-D-glucose hydrochloride, Chitosamine hydrochloride
Empirical Formula (Hill Notation):
C6H13NO5 · HCl
CAS Number:
Molecular Weight:
215.63
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing