Skip to Content
BE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gluconolactone
Synonym(s):
Gluconolactone, D-(+)-Gluconic acid δ-lactone, δ-Gluconolactone, 1,2,3,4,5-Pentahydroxycaproic acid δ-lactone, D-(+)-Dextronic acid δ-lactone
Empirical Formula (Hill Notation):
C
6
H
10
O
6
CAS Number:
90-80-2
Molecular Weight:
178.14
EC Number:
202-016-5
Products
Product Category
carbohydrates (1)
Brand
Millipore (1)
Sigma-Aldrich (1)
Biological Source
Color
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
347425
Gluconolactone,
An oxidation product of glucose by glucose oxidase.
Expand
View Pricing
1294150
Gluconolactone,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
G2164
Gluconolactone,
99.0-101.0%, meets USP testing specifications
Expand
View Pricing