Skip to Content
BE
EN
Back
Gitoxin
Synonym(s):
Anhydrogitalin, Bigitalin, Pseudodigitoxin
Empirical Formula (Hill Notation):
C
41
H
64
O
14
CAS Number:
4562-36-1
Molecular Weight:
780.94
Products
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
G0300000
Gitoxin,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing