![Ginsenoside Compound K ≥96% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/202/580/a2394a13-83de-43c4-a47f-4fef0e8b9a01/640/a2394a13-83de-43c4-a47f-4fef0e8b9a01.png)
Ginsenoside Compound K
![Ginsenoside Compound K ≥96% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/202/580/a2394a13-83de-43c4-a47f-4fef0e8b9a01/640/a2394a13-83de-43c4-a47f-4fef0e8b9a01.png)
Synonym(s):
(3B,12B)-3,12-Dihydroxydammar-24-en-20-yl β-D-glucopyranoside, 20(S)-Ginsenoside Ck, Ginsenoside K, Protopanaxadiol 20-O-glucoside
Empirical Formula (Hill Notation):
C36H62O8
CAS Number:
Molecular Weight:
622.87
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing