Skip to Content
Merck
Genistein primary reference standard

Genistein

Genistein primary reference standard
Synonym(s):
4′,5,7-Trihydroxyisoflavone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, Genistein
Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique