![Gabapentin Related Compound D Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/126/848/b55ec3e5-c35f-4f87-8e1f-5e88b135fe8e/640/b55ec3e5-c35f-4f87-8e1f-5e88b135fe8e.png)
Gabapentin Related Compound D
![Gabapentin Related Compound D Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/126/848/b55ec3e5-c35f-4f87-8e1f-5e88b135fe8e/640/b55ec3e5-c35f-4f87-8e1f-5e88b135fe8e.png)
Synonym(s):
1-[(3-Oxo-2-azaspiro[4.5]dec-2-yl)methyl]cyclohexaneacetic acid
Empirical Formula (Hill Notation):
C18H29NO3
CAS Number:
Molecular Weight:
307.43
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2