![G-418 Solution solution, =98% (TLC), suitable for transfection](/deepweb/assets/sigmaaldrich/product/structures/296/709/19ac2b23-4ba0-4722-a20a-8261dec4be46/640/19ac2b23-4ba0-4722-a20a-8261dec4be46.png)
G-418 Solution
![G-418 Solution solution, =98% (TLC), suitable for transfection](/deepweb/assets/sigmaaldrich/product/structures/296/709/19ac2b23-4ba0-4722-a20a-8261dec4be46/640/19ac2b23-4ba0-4722-a20a-8261dec4be46.png)
Synonym(s):
G 418 disulfate salt solution, Antibiotic G418
Empirical Formula (Hill Notation):
C20H40N4O10 · 2H2SO4
CAS Number:
Molecular Weight:
692.71
Products
Product Category
Formula Weight
Manufacturer
Physical Form
Activity Spectrum
Quality Segment
Mode of Action
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing