![Frangulin A phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/215/849/a9bbeb10-01ee-4524-841b-06b9f9e1bb84/640/a9bbeb10-01ee-4524-841b-06b9f9e1bb84.png)
Frangulin A
![Frangulin A phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/215/849/a9bbeb10-01ee-4524-841b-06b9f9e1bb84/640/a9bbeb10-01ee-4524-841b-06b9f9e1bb84.png)
Synonym(s):
3-[(6-Deoxy-α-L-mannopyranosyl)oxy]-1,8-dihydroxy-6-methylanthraquinone, Emodin-L-rhamnoside, Franguloside, Rhamnoxanthin
Empirical Formula (Hill Notation):
C21H20O9
CAS Number:
Molecular Weight:
416.38
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing