Skip to Content
BE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Fractogel
®
EMD DMAE (M)
Products
Product Category
columns (2)
Brand
Millipore (3)
Physical Form
Product Line
Separation Technique
ID (mm)
Volume (ml)
Quality Segment
Application
Housing Material
Particle Size (µm)
Sterility
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
1.16884
Fractogel
®
EMD DMAE (M),
Expand
View Pricing
1.25080
Fractogel
®
EMD DMAE (M),
MiniChrom Fractogel
®
DMAE (M), volume 5 mL, prepacked column
Expand
View Pricing
125059
Fractogel
®
EMD DMAE (M),
MiniChrom Fractogel
®
DMAE (M), volume 1 mL, prepacked column
Expand
View Pricing