Forchlorfenuron
Synonym(s):
N-(2-Chloro-4-pyridyl)-N′-phenylurea, 4-CPPU, CPPU
Empirical Formula (Hill Notation):
C12H10ClN3O
CAS Number:
Molecular Weight:
247.68
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2