Fluorouracil Related Compound B
Synonym(s):
Isobarbituric acid, 2,4,5-Trihydroxypyrimidine, 5-Hydroxyuracil
Empirical Formula (Hill Notation):
C4H4N2O3
CAS Number:
Molecular Weight:
128.09
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2