Fenbendazole Related Compound B
Synonym(s):
Methyl (5-chloro-1H-benzimidazol-2-yl)carbamate
Empirical Formula (Hill Notation):
C9H8ClN3O2
CAS Number:
Molecular Weight:
225.63
Products
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2