![Ethynodiol diacetate British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/241/699/c7f8f52f-dc18-43bb-b529-13b65117bb1e/640/c7f8f52f-dc18-43bb-b529-13b65117bb1e.png)
Ethynodiol diacetate
![Ethynodiol diacetate British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/241/699/c7f8f52f-dc18-43bb-b529-13b65117bb1e/640/c7f8f52f-dc18-43bb-b529-13b65117bb1e.png)
Synonym(s):
(3β, 17α)-19-Norpregn-4-en-20-yne-3,17-diol diacetate, 3β,17β-Diacetoxy-17α-ethynyl-4-estrene, Etynodiol acetate
Empirical Formula (Hill Notation):
C24H32O4
CAS Number:
Molecular Weight:
384.51
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3