Skip to Content
Merck
(−)-Ethyl L-lactate analytical standard

(−)-Ethyl L-lactate

(−)-Ethyl L-lactate analytical standard
Synonym(s):
(−)-Ethyl (S)-2-hydroxypropionate, (S)-(−)-2-Hydroxypropionic acid ethyl ester, L-Lactic acid ethyl ester
Empirical Formula (Hill Notation):
C5H10O3
CAS Number:
Molecular Weight:
118.13
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Reference Material Type
Special Grade
Showing 1-3 of 3