Skip to Content
Merck
Ethylenediamine-N,N′-disuccinic acid trisodium salt solution ~35% in H2O

Ethylenediamine-N,N′-disuccinic acid trisodium salt solution

Ethylenediamine-N,N′-disuccinic acid trisodium salt solution ~35% in H2O
Synonym(s):
N,N′-Ethylenedi-(aspartic acid) trisodium salt, EDDS-Na3, Trisodium ethylenediamine-N,N′-disuccinate
Empirical Formula (Hill Notation):
C10H13N2Na3O8
Molecular Weight:
358.19

Products

Product Category
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing