Skip to Content
Merck
Estradiol hemihydrate British Pharmacopoeia (BP) Assay Standard

Estradiol hemihydrate

Estradiol hemihydrate British Pharmacopoeia (BP) Assay Standard
Synonym(s):
β-Estradiol hemihydrate, 1,3,5-Estratriene-3,17β-diol, 3,17β-Dihydroxy-1,3,5(10)-estratriene
Empirical Formula (Hill Notation):
C18H24O2 · 0.5H2O
CAS Number:
Molecular Weight:
281.39
EC Number:

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade