Skip to Content
Merck
Esculin hydrate ≥98.0% (HPLC)

Esculin hydrate

Esculin hydrate ≥98.0% (HPLC)
Empirical Formula (Hill Notation):
C15H16O9 · xH2O
CAS Number:
Molecular Weight:
340.28 (anhydrous basis)

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-2 of 2
Compare
Product Number
Description
Pricing