Skip to Content
BE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Reference 2 lower part
Products
Color
Manufacturer
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
EP4920611007
Eppendorf
®
Reference 2 lower part,
single-channel, lower part with separate serial number, turquoise, size 1-10 mL
Expand
View Pricing
EP4922602005
Eppendorf
®
Reference 2 lower part,
12-channel, lower part with separate serial number, medium gray, size 0.5-10 μL
Expand
View Pricing
EP4922605004
Eppendorf
®
Reference 2 lower part,
8-channel, lower part with separate serial number, orange, size 30-300 μL
Expand
View Pricing
EP4920608006
Eppendorf
®
Reference 2 lower part,
single-channel, lower part with separate serial number, blue, size 100-1000 μL
Expand
View Pricing
EP4920610000
Eppendorf
®
Reference 2 lower part,
single-channel, lower part with separate serial number, violet, size 0.5-5 μL
Expand
View Pricing
EP4922604008
Eppendorf
®
Reference 2 lower part,
12-channel, lower part with separate serial number, yellow, size 10-100 μL
Expand
View Pricing
EP4920605007
Eppendorf
®
Reference 2 lower part,
single-channel, lower part with separate serial number, yellow, size 10-100 μL
Expand
View Pricing
EP4920609002
Eppendorf
®
Reference 2 lower part,
single-channel, lower part with separate serial number, red, size 0.25-2.5 mL
Expand
View Pricing
EP4922606000
Eppendorf
®
Reference 2 lower part,
12-channel, lower part with separate serial number, orange, size 30-300 μL
Expand
View Pricing
EP4922601009
Eppendorf
®
Reference 2 lower part,
8-channel, lower part with separate serial number, medium gray, size 0.5-10 μL
Expand
View Pricing
EP4922603001
Eppendorf
®
Reference 2 lower part,
8-channel, lower part with separate serial number, yellow, size 10-100 μL
Expand
View Pricing
EP4920602008
Eppendorf
®
Reference 2 lower part,
single-channel, lower part with separate serial number, medium gray, size 0.5-10 μL
Expand
View Pricing
EP4920601001
Eppendorf
®
Reference 2 lower part,
single-channel, lower part with separate serial number, dark gray, size 0.1-2.5 μL
Expand
View Pricing
EP4920603004
Eppendorf
®
Reference 2 lower part,
single-channel, lower part with separate serial number, light gray, size 2-20 μL
Expand
View Pricing
EP4920604000
Eppendorf
®
Reference 2 lower part,
single-channel, lower part with separate serial number, yellow, size 2-20 μL
Expand
View Pricing
EP4920607000
Eppendorf
®
Reference 2 lower part,
single-channel, lower part with separate serial number, yellow, size 30-300 μL
Expand
View Pricing