Skip to Content
BE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Reference 2 Ejector sleeve
Products
Manufacturer
Filter Products
Showing 1-19 of 19
Compare
Product Number
Description
Pricing
EP4920615002
Eppendorf
®
Reference 2 Ejector sleeve,
variable, size 100 μL, color code: yellow
Expand
View Pricing
EP4920613000
Eppendorf
®
Reference 2 Ejector sleeve,
variable, size 10 μL, color code: medium gray or yellow
Expand
View Pricing
EP4921601005
Eppendorf
®
Reference 2 Ejector sleeve,
fixed, size 1 μL, color code: dark gray
Expand
View Pricing
EP4920620006
Eppendorf
®
Reference 2 Ejector sleeve,
variable, size 5 mL, color code: violet
Expand
View Pricing
EP4920616009
Eppendorf
®
Reference 2 Ejector sleeve,
variable, size 200 μL, color code: yellow
Expand
View Pricing
EP4921606007
Eppendorf
®
Reference 2 Ejector sleeve,
fixed, size 200 μL, color code: blue
Expand
View Pricing
EP4920619008
Eppendorf
®
Reference 2 Ejector sleeve,
variable, size 2.5 mL, color code: red
Expand
View Pricing
EP4920621002
Eppendorf
®
Reference 2 Ejector sleeve,
variable, size 10 mL, color code: turquoise
Expand
View Pricing
EP4920618001
Eppendorf
®
Reference 2 Ejector sleeve,
variable, size 1000 μL, color code: blue
Expand
View Pricing
EP4920614006
Eppendorf
®
Reference 2 Ejector sleeve,
variable, size 20 μL, color code: light gray or yellow
Expand
View Pricing
EP4921609006
Eppendorf
®
Reference 2 Ejector sleeve,
fixed, size 2 mL, color code: red
Expand
View Pricing
EP4921604004
Eppendorf
®
Reference 2 Ejector sleeve,
fixed, size 25 μL, color code: yellow
Expand
View Pricing
EP4921602001
Eppendorf
®
Reference 2 Ejector sleeve,
fixed, size 2 μL, color code: dark gray
Expand
View Pricing
EP4921607003
Eppendorf
®
Reference 2 Ejector sleeve,
fixed, size 250 μL, color code: blue
Expand
View Pricing
EP4921603008
Eppendorf
®
Reference 2 Ejector sleeve,
fixed, size 5 μL, color code: medium gray
Expand
View Pricing
EP4921605000
Eppendorf
®
Reference 2 Ejector sleeve,
fixed, size 50 μL, color code: yellow
Expand
View Pricing
EP4921608000
Eppendorf
®
Reference 2 Ejector sleeve,
fixed, size 500 μL, color code: blue
Expand
View Pricing
EP4920612003
Eppendorf
®
Reference 2 Ejector sleeve,
variable, size 2.5 μL, color code: dark gray
Expand
View Pricing
EP4920617005
Eppendorf
®
Reference 2 Ejector sleeve,
variable, size 300 μL, color code: orange
Expand
View Pricing