Skip to Content
Merck
Emetine hydrochloride European Pharmacopoeia (EP) Reference Standard

Emetine hydrochloride

Emetine hydrochloride European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Emetine dihydrochloride, 6′,7′,10,11-Tetramethoxyemetan dihydrochloride
Empirical Formula (Hill Notation):
C29H40N2O4 · 2HCl
CAS Number:
Molecular Weight:
553.56

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade