Skip to Content
Merck
Embelin phyproof® Reference Substance

Embelin

Embelin phyproof® Reference Substance
Synonym(s):
2,5-Dihydroxy-3-undecyl-2,5-cyclohexadiene-1,4-dione, Embelic acid, Emberine
Empirical Formula (Hill Notation):
C17H26O4
CAS Number:
Molecular Weight:
294.39

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Activity Spectrum
Format
Research Area
Application
Mode of Action
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing