E. coli WDCM 00202 Vitroids
Synonym(s):
Escherichia coli WDCM 00202 Vitroids
Products
Product Category
Brand
Manufacturer
Physical Form
Agency
Quality Segment
Application
Microorganism Suitability
Package Size
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing